missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012494
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012494 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012494 Supplier Merck Emd Millipore Supplier No. PHR1632500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 112529-15-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H20N2O3 S · HCl
MDL Number MFCD04975446
Melting Point 190°C to 195.5°C
Synonym Pioglitazone hydrochloride; 5-[4-[2-(5-Ethyl-2-pyridinyl)ethoxy]benzyl]thiazolidine-2,4-dione hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 392.90
Grade Certified Reference Material