missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone Hydrochloride 98.0+%, TCI America™

Catalog No. P19015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19015G 5 g
P190125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19015G Supplier TCI America Supplier No. P19015G

Chemical Identifiers

CAS 112529-15-4
Molecular Formula C19H21ClN2O3S
Molecular Weight (g/mol) 392.898
MDL Number MFCD04975446
InChI Key GHUUBYQTCDQWRA-UHFFFAOYSA-N
Synonym 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione Hydrochloride
PubChem CID 60560
IUPAC Name 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
SMILES CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl

Specifications

Physical Form Powder/Crystals
Color White
Melting Point 191°C
Quantity 5 g
Formula Weight 392.90
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material Pioglitazone Hydrochloride