accessibility menu, dialog, popup

UserName

OHOOBr

CAS RN 1113-59-3

IUPAC Name: 3-bromo-2-oxopropanoic acid
Synonyms: bromopyruvic acid
Molecular Weight (g/mol): 166.96
Molecular Formula: C3H3BrO3
InChi Key: PRRZDZJYSJLDBS-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)CBr
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)