missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Hexokinase II Inhibitor II, 3-BP

Catalog No. 376817100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
376817100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 376817100MG Supplier MilliporeSigma™ Supplier No. 376817100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Hexokinase II Inhibitor II, 3-BP, also referenced under CAS 1113-59-3, controls the biological activity of Hexokinase II. This small molecule/inhibitor is primarily used for Cell Structure applications.

A cell-permeable pyruvic acid derivative that acts as an inhibitor against hexokinase II-mediated glycolysis and effectively suppress tumor survival both in cultures in vitro and in mice and rats in vivo .

Specifications

Color White
Description 97% by Titration
Product Type Inhibitor
CAS 1113-59-3
Molecular Formula C3H3BrO3
Formula Weight 166.96g/mol
Solubility DMSO or dH2O
Form Crystalline Solid
Quantity 100 mg