missing translation for 'onlineSavingsMsg'
Learn More

Bromopyruvic acid, 97%

Catalog No. AAL0072018
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0072018 50 g
AAL0072004 2 g
AAL0072009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0072018 Supplier Thermo Scientific Chemicals Supplier No. L0072018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1113-59-3 | C3H3BrO3 | 166.958 g/mol

Bromopyruvic acid is used as an affinity label for cysteine residues and a potential anti-cancer agent. It is involved in the synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids and substituted pyranones. It plays a vital role as a cross-linker between nucleic acids and proteins. It is used as an anticancer agent for lung tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromopyruvic acid is used as an affinity label for cysteine residues and a potential anti-cancer agent. It is involved in the synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids and substituted pyranones. It plays a vital role as a cross-linker between nucleic acids and proteins. It is used as an anticancer agent for lung tumors.

Solubility
Soluble in water, dimethyl sulfoxide, ethyl acetate and water.

Notes
Moisture and air sensitive. Store in cool place. Incompatible with strong oxidizing agents and reducing agents.

Chemical Identifiers

CAS 1113-59-3
Molecular Formula C3H3BrO3
Molecular Weight (g/mol) 166.958
MDL Number MFCD00002587
InChI Key PRRZDZJYSJLDBS-UHFFFAOYSA-N
Synonym 3-bromopyruvic acid, bromopyruvic acid, bromopyruvate, 3-bromopyruvate, 3-bromo-2-oxopropionic acid, bromopyruvicacid, pyruvic acid, bromo, propanoic acid, 3-bromo-2-oxo, unii-63jmv04grk, .beta.-bromopyruvic acid
PubChem CID 70684
IUPAC Name 3-bromo-2-oxopropanoic acid
SMILES C(C(=O)C(=O)O)Br

Specifications

Melting Point 75°C to 80°C
Quantity 50 g
UN Number UN3261
Beilstein 1746786
Solubility Information Soluble in water,dimethyl sulfoxide,ethyl acetate and water.
Formula Weight 166.96
Percent Purity 97%
Chemical Name or Material Bromopyruvic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 214-206-5
RTECSNumber UZ0840000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only