accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 111-61-5

IUPAC Name: ethyl octadecanoate
Synonyms: ethyl stearate
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: MVLVMROFTAUDAG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)