missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Stearate 96.0+%, TCI America™

Catalog No. S007925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S007925G 25 g
S0079250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S007925G Supplier TCI America Supplier No. S007925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-61-5
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
MDL Number MFCD00009006
InChI Key MVLVMROFTAUDAG-UHFFFAOYSA-N
Synonym ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate
PubChem CID 8122
ChEBI CHEBI:84936
IUPAC Name ethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Color White-Yellow
Boiling Point 199°C
Quantity 25 g
Formula Weight 312.54
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl Stearate
EINECSNumber (2)-0798
RTECSNumber WI3600000
TSCA Yes