accessibility menu, dialog, popup

UserName

HONHOH

CAS RN 111-42-2

IUPAC Name: 2-[(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: diethanolamine diaethanolamin dabco deoa-LF diethanolamine (substance) N,N'-iminodiethanol dihydroxyethyl tallowamine oxide diethyolamine niax deoa-LF H2dea N,N-diethanolamine
Molecular Weight (g/mol): 105.14
Molecular Formula: C4H11NO2
InChi Key: ZBCBWPMODOFKDW-UHFFFAOYSA-N
SMILES: OCCNCCO
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (7)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (7)
  • (1)
  • (2)
  • (4)
Boiling Point 
  • (7)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (7)
  • (2)
  • (1)
  • (7)
  • (4)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (4)
  • (7)
  • (3)
  • (1)
  • (2)
  • (4)
  • (7)
  • (2)