missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine 99.0+%, TCI America™

Catalog No. I000825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I000825G 25 g
I0008500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I000825G Supplier TCI America Supplier No. I000825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

Color White-Yellow
pH 11.0 (0.1N H2O soln.)
Boiling Point 269°C
Quantity 25 g
Formula Weight 105.14
Percent Purity ≥99.0% (GC,T)
Grade Not Available
Physical Form Crystalline Lumps
Chemical Name or Material Diethanolamine
EINECSNumber (2)-0302&(2)-0354
RTECSNumber KL2975000
TSCA Yes