missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019158
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019158 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019158 Supplier Merck Emd Millipore Supplier No. 169575MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diethanolamine (DEA) is an organic chemical compound, which comprises of a secondary amine and a dialcohol. It mainly acts as a weak base. DEA can be used in preparing long-chain fatty acids, namely diethanolamides and diethanolamine salts, which find application in liquid laundry and dishwashing detergents, cosmetics, shampoos, hair conditioners, textile processing, and also in pharmaceutical industries. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 111-42-2
Ignition Point 689°F
Vapor Pressure <0.98 atm (100°C)
Vapor Density 3.6 (vs. air)
Density 1.097 g/mL (at 25°C)
Flash Point 138°C
Physical Form Neat
Linear Formula HN(CH2 CH2 OH)2
MDL Number MFCD00002843
Melting Point 28°C
Boiling Point 217°C (150 mmHg)
Synonym 2,2′-Iminodiethanol; Bis(2-hydroxyethyl)amine
Type Analytical Standard
Quantity 5 mL
Formula Weight 105.14
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less