CAS RN 110-44-1
Sorbic acid, 99%
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N Synonym: sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
phyproof™ Sorbic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
2,4-Hexadienoic acid, 99%
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N Synonym: sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid 99.0+%, TCI America™
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N Synonym: sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid Standard, 250 ppm C6H8O2, Ricca Chemical
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid Standard, 300 ppm C6H8O2, Ricca Chemical
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid Standard, 100 ppm C6H8O2, Ricca Chemical
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic acid, free acid, ≥99%, MP Biomedicals™
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N Synonym: sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid 99.0+%, TCI America™
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N Synonym: sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid PubChem CID: 643460 ChEBI: CHEBI:38358 IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Medisca Sorbic Acid NF (Powder)
Sorbic Acid NF is a free-flowing, white, crystalline powder, having a characteristic odor. Alternative name(s): 2,4-Hexadienoic acid
Sorbic Acid, Powder, FCC, 99-101%, Spectrum™ Chemical
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid, Powder, NF, 99-101%, Spectrum™ Chemical
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O
Sorbic Acid, Powder, NF, 99-101%, Spectrum™ Chemical
CAS: 110-44-1 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.13 MDL Number: MFCD00002703 InChI Key: WSWCOQWTEOXDQX-MQQKCMAXSA-N IUPAC Name: (2E,4E)-hexa-2,4-dienoic acid SMILES: C\C=C\C=C\C(O)=O