missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Sorbic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002408
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002408 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002408 Supplier Merck Emd Millipore Supplier No. PHL80020100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 110-44-1
Product Line phyproof
Vapor Pressure 0.01 mmHg (20°C)
Flash Point 127°C - closed cup
Linear Formula CH3 CH=CHCH=CHCOOH
MDL Number MFCD00002703
Melting Point 132°C to 135°C
Synonym alpha-trans-gamma-trans-Sorbic acid; (E,E)-1,3-Pentadiene-1-carboxylic acid; 2,4-Hexadienoic acid; 2 E,4 E-Hexadienoic acid; trans,trans-2,4-Hexadienoic acid; C6:2 n-2,4; NSC 35405; NSC 49103; NSC 50268
Type Reference Substance
Quantity 100 mg
Formula Weight 112.13
Percent Purity ≥98% (HPLC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG