missing translation for 'onlineSavingsMsg'
Learn More

Medisca Sorbic Acid NF (Powder)

Catalog No. 14185454
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185454 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185454 Supplier Medisca Supplier No. 077105
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sorbic Acid NF is a free-flowing, white, crystalline powder, having a characteristic odor. Alternative name(s): 2,4-Hexadienoic acid

Sorbic Acid NF is a free-flowing, white, crystalline powder, having a characteristic odor.

Alternative name(s): 2,4-Hexadienoic acid

Specifications

Chemical Name or Material Sorbic Acid NF (Powder)
Packaging Jar, 150mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 110-44-1
Type Drug Standard
Molecular Weight (g/mol) 112.127
Quantity 100 g
Percent Purity 98 to 102 %
Grade NF