accessibility menu, dialog, popup

UserName

CAS RN 108964-32-5

IUPAC Name: (acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}[2-(5-{[(acetyloxy)methoxy]carbonyl}-1,3-oxazol-2-yl)-4-(2-{2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]-5-methylphenoxy}ethoxy)-1-benzofuran-5-yl]amino)acetate
Synonyms: fura-2-acetoxymethyl ester
Molecular Weight (g/mol): 1001.86
Molecular Formula: C44H47N3O24
InChi Key: UTBYGIDPBQOHNT-UHFFFAOYSA-N
SMILES: CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)C1=CC=C(C)C=C1OCCOC1=C2C=C(OC2=CC=C1N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C1=NC=C(O1)C(=O)OCOC(C)=O
Form 
  • (1)
  • (1)
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results