missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ FURA 2/AM

Catalog No. 3449051MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3449051MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3449051MG Supplier MilliporeSigma™ Supplier No. 3449051MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell-permeable ester form of FURA 2

  • Cell-permeable ester form of FURA 2
  • PLURONIC™ F-127, PROTEIN GRADE™ Detergent is a mild, low-toxicity detergent that can aid in the solubilization of FURA 2/AM

Specifications

CAS 108964-32-5
Color Yellow
Content And Storage Store at −20°C
Formula Weight 1001.9g/mol
Packaging Type Packaged under inert gas
Purity ≥95% by HPLC
Quantity 1 mg
Synonym 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2′-amino-5′-methylphenoxy)-ethane-N,N,N′N′-tetraacetic Acid Pentaacetoxymethyl Ester
Target Ratiometric calcium indicator
Form Solid