missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ FURA 2/AM

Catalog No. 3449061ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3449061ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3449061ML Supplier MilliporeSigma™ Supplier No. 3449061ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1 mM solution of FURA 2/AM

A 1 mM solution of FURA 2/AM in anhydrous DMSO

Specifications

CAS 108964-32-5
Content And Storage Store at −20°C
Formula Weight 1001.9g/mol
Formulation A 1 mM solution of FURA 2/AM in anhydrous DMSO
Packaging Type Packaged under inert gas
Purity ≥95% by HPLC
Quantity 1 mL
Form Liquid