accessibility menu, dialog, popup

UserName

CAS RN 1083-57-4

IUPAC Name: N-(4-ethoxyphenyl)-3-hydroxybutanamide
Synonyms: bucetin betadid bucetine bucetina β-oxybuttersaeure-p-phenetidid 布西丁 bucetinum
Molecular Weight (g/mol): 223.27
Molecular Formula: C12H17NO3
InChi Key: LIAWQASKBFCRNR-UHFFFAOYNA-N
SMILES: CCOC1=CC=C(NC(=O)CC(C)O)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results