missing translation for 'onlineSavingsMsg'
Learn More

Bucetin, MilliporeSigma™ Supelco™

Catalog No. 111012430
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012430 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012430 Supplier Merck Emd Millipore Supplier No. B402710G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 1083-57-4
Physical Form Neat
Linear Formula C12H17 NO3
MDL Number MFCD00021906
Synonym 3-Hydroxy-p-butyrophenetidide; N-(4-Ethoxyphenyl)-3-hydroxybutanamide
Type Analytical Standard
Quantity 10 g
Formula Weight 223.27
Grade Analytical Standard