missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-p-butyrophenetidine 98.0+%, TCI America™

Catalog No. H037125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H037125G Supplier TCI America Supplier No. H037125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1083-57-4
Molecular Formula C12H17NO3
Molecular Weight (g/mol) 223.27
MDL Number MFCD00021906
InChI Key LIAWQASKBFCRNR-UHFFFAOYNA-N
Synonym 4′C-Ethoxy-3-hydroxybutyranilide, N-(3-Hydroxybutyryl)-p-phenetidine
PubChem CID 14130
IUPAC Name N-(4-ethoxyphenyl)-3-hydroxybutanamide
SMILES CCOC1=CC=C(NC(=O)CC(C)O)C=C1

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 223.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-p-butyrophenetidine
EINECSNumber (3)-0696
RTECSNumber EU0900000
TSCA Yes