accessibility menu, dialog, popup

UserName

CAS RN 108-73-6

IUPAC Name: benzene-1,3,5-triol
Synonyms: phloroglucinol floroglucin s-trihydroxybenzene trihydroxy benzene floroglucinol benzene-s-triol dilospan S spassirex lafon brand of phloroglucinol spasfon-lyoc
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: QCDYQQDYXPDABM-UHFFFAOYSA-N
SMILES: OC1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)