missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol, 99+%, anhydrous

Catalog No. AC241765000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241765000 500 g Glass bottle
AC241760250 25 g Glass bottle
AC241761000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC241765000 Supplier Thermo Scientific Chemicals Supplier No. 241765000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-73-6 | C6H6O3 | 126.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-73-6
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.1
InChI Key QCDYQQDYXPDABM-UHFFFAOYSA-N
Synonym phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy
PubChem CID 359
ChEBI CHEBI:16204
IUPAC Name benzene-1,3,5-triol
SMILES C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 215.0°C to 218.0°C
Color White to Beige
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula (OH)3C6H3
Quantity 500 g
Fieser 01,47; 06,147
Merck Index 15,7439
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone ethanol, ether and pyridine
Formula Weight 126.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 203-611-2

RUO – Research Use Only