missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol Anhydrous 99.0+%, TCI America™

Catalog No. P024925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024925G 25 g
P0249250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P024925G Supplier TCI America Supplier No. P024925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-73-6
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00002286
InChI Key QCDYQQDYXPDABM-UHFFFAOYSA-N
Synonym phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy
PubChem CID 359
ChEBI CHEBI:16204
IUPAC Name benzene-1,3,5-triol
SMILES C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 218°C
Quantity 25 g
Formula Weight 126.11
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol Anhydrous
EINECSNumber (3)-0554
RTECSNumber SY1050000
TSCA Yes