accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 106-70-7

IUPAC Name: methyl hexanoate
Synonyms: methyl caproate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: NUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OC
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)