missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl hexanoate, 99%, analytical standard for GC, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | Methyl hexanoate |
| CAS | 106-70-7 |
| Densité | 0.8840g/mL |
| Point d’ébullition | 151.0°C |
| Point d’éclair | 42°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 98.5% min. (GC) |
| Emballage | Glass bottle |
| Formule moléculaire | C7H14O2 |
| Indice de réfraction | 1.4047 to 1.4067 |
| Afficher plus de résultats |
RUO – Research Use Only