missing translation for 'onlineSavingsMsg'
Learn More

Methyl hexanoate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020199
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020199 1 mL
111018794 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020199 Supplier Merck Emd Millipore Supplier No. 215991MLF

Specifications

CAS 106-70-7
Density 0.884 g/mL (at 20°C)
Flash Point 43°C
Physical Form Neat
Linear Formula CH3(CH2)4 COOCH3
MDL Number MFCD00009510
Synonym Caproic acid methyl ester; Hexanoic acid methyl ester; Methyl caproate
Type Analytical Standard
Quantity 1 mL
Formula Weight 130.18
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less