missing translation for 'onlineSavingsMsg'
Learn More

Methyl hexanoate, 99%, analytical standard for GC, Thermo Scientific™

Catalog No. AC414420050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414420050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414420050 Supplier Thermo Scientific Chemicals Supplier No. 414420050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Methyl hexanoate
Name Note 98%
Acidity 1mg KOH/g max.
CAS 106-70-7
Density 0.8840g/mL
Boiling Point 151.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H14O2
Refractive Index 1.4047 to 1.4067
Linear Formula CH3(CH2)4COOCH3
Quantity 5g
MDL Number MFCD00009510
Specific Gravity 0.884
Synonym methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
Solubility Information Solubility in water: insoluble
InChI Key NUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILES CCCCCC(=O)OC
IUPAC Name methyl hexanoate
Molecular Weight (g/mol) 130.19
PubChem CID 7824
Viscosity <30 mPa.s (20°C)
ChEBI CHEBI:77322
Formula Weight 130.18
Percent Purity 99%
Show More Show Less

RUO – Research Use Only