accessibility menu, dialog, popup

UserName

CAS RN 10478-33-8

HCNOO

CAS RN 10478-33-8

IUPAC Name: 1-(prop-2-yn-1-yl)pyrrolidine-2,5-dione
Synonyms: 1-(prop-2-yn-1-yl)pyrrolidine-2,5-dione
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: DOTAQRZGKATJIC-UHFFFAOYSA-N
SMILES: O=C1CCC(=O)N1CC#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results