missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Propynyl)succinimide 98.0+%, TCI America™

Catalog No. P2191200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2191200MG 200 mg
P21911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2191200MG Supplier TCI America Supplier No. P2191200MG
Only null left

Chemical Identifiers

CAS 10478-33-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD06740819
InChI Key DOTAQRZGKATJIC-UHFFFAOYSA-N
Synonym N-Propargylsuccinimide
PubChem CID 4737238
IUPAC Name 1-(prop-2-yn-1-yl)pyrrolidine-2,5-dione
SMILES O=C1CCC(=O)N1CC#C

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 237°C
Quantity 200 mg
Formula Weight 137.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Propynyl)succinimide