accessibility menu, dialog, popup

UserName

CAS RN 104-85-8

IUPAC Name: 4-methylbenzonitrile
Synonyms: 4-methylbenzonitrile
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: VCZNNAKNUVJVGX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C#N
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)