accessibility menu, dialog, popup

UserName

CAS RN 10284-63-6

IUPAC Name: (1R,2S,4S,5S)-6-methoxycyclohexane-1,2,3,4,5-pentol
Synonyms: methylinositol
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: DSCFFEYYQKSRSV-FEPQRWDDSA-N
SMILES: COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)