missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Pinitol, 95%

Catalog No. AAH56648MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56648MD 250 mg
AAH5664803 1 g
AAH5664806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH56648MD Supplier Thermo Scientific Chemicals Supplier No. H56648MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10284-63-6 | C7H14O6 | 194.18 g/mol

It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and  has shown antidiabetic properties in mice. It is  believed to be a salt stress regulator in a wide range of plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and  has shown antidiabetic properties in mice. It is  believed to be a salt stress regulator in a wide range of plants.

Solubility
Soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 10284-63-6
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00216659
InChI Key DSCFFEYYQKSRSV-FEPQRWDDSA-N
Synonym d-pinitol, pinitol, 3-o-methyl-d-chiro-inositol, d-+-pinitol, inzitol, pinit, +-pinitol, methylinositol, sennitol, d-ononitol
PubChem CID 164619
IUPAC Name (1S,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol
SMILES COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O

Specifications

Melting Point 178°C to 185°C
Quantity 250 mg
Merck Index 14,10361
Solubility Information Soluble in water.
Optical Rotation +60° (c=1 in Water)
Formula Weight 194.18
Percent Purity 95%
Chemical Name or Material D-Pinitol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only