missing translation for 'onlineSavingsMsg'
Learn More

D-Pinitol 98.0+%, TCI America™

Catalog No. P22191G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22191G 1 g
P2219100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22191G Supplier TCI America Supplier No. P22191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10284-63-6
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00216659
InChI Key DSCFFEYYQKSRSV-FEPQRWDDSA-N
Synonym d-pinitol, pinitol, 3-o-methyl-d-chiro-inositol, d-+-pinitol, inzitol, pinit, +-pinitol, methylinositol, sennitol, d-ononitol
PubChem CID 164619
IUPAC Name (1R,2S,4S,5S)-6-methoxycyclohexane-1,2,3,4,5-pentol
SMILES COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Formula Weight 194.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-Pinitol