accessibility menu, dialog, popup

UserName

CAS RN 100900-07-0

IUPAC Name: sodium 5-[(2-aminoethyl)amino]naphthalene-1-sulfonate
Synonyms: sodium 5-[(2-aminoethyl)amino]naphthalene-1-sulfonate
Molecular Weight (g/mol): 288.3
Molecular Formula: C12H13N2NaO3S
InChi Key: HGWRACRQRUQQGH-UHFFFAOYSA-M
SMILES: [Na+].NCCNC1=C2C=CC=C(C2=CC=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)