missing translation for 'onlineSavingsMsg'
Learn More

Sodium 5-(2-Aminoethylamino)-1-naphthalenesulfonate Hydrate 98.0+%, TCI America™

Catalog No. S0891100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0891100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0891100MG Supplier TCI America Supplier No. S0891100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100900-07-0
Molecular Formula C12H13N2NaO3S
Molecular Weight (g/mol) 288.297
MDL Number MFCD00051474
InChI Key HGWRACRQRUQQGH-UHFFFAOYSA-M
Synonym sodium 5-2-aminoethyl amino naphthalene-1-sulfonate, 5-2-aminoethylamino-1-naphthalenesulfonic acid sodium salt, sodium 5-2-aminoethylamino naphthalene-1-sulfonate, n-aminoethyl-5-naphthylamine-1-sulfonic acid sodium salt, 5-2-aminoethylamino-1-naphthalenesulphonic acid sodium salt, 1,5-edans, sodium salt, sodium 5-2-aminoethylamino-1-naphthalenesulfonate, sodium n-2-aminoethyl-5-naphthylamine-1-sulfonate, sodium5-2-aminoethylamino-1-naphthalenesulfonatehydrate
PubChem CID 4116252
IUPAC Name sodium;5-(2-aminoethylamino)naphthalene-1-sulfonate
SMILES C1=CC2=C(C=CC=C2S(=O)(=O)[O-])C(=C1)NCCN.[Na+]

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 288.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 5-(2-Aminoethylamino)-1-naphthalenesulfonate Hydrate