missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt, 97%

Catalog No. AAL06555MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06555MD 250 mg
AAL0655503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL06555MD Supplier Thermo Scientific Chemicals Supplier No. L06555MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100900-07-0 | C12H13N2NaO3S | 288.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100900-07-0
Molecular Formula C12H13N2NaO3S
Molecular Weight (g/mol) 288.297
MDL Number MFCD00051474
InChI Key HGWRACRQRUQQGH-UHFFFAOYSA-M
Synonym sodium 5-2-aminoethyl amino naphthalene-1-sulfonate, 5-2-aminoethylamino-1-naphthalenesulfonic acid sodium salt, sodium 5-2-aminoethylamino naphthalene-1-sulfonate, n-aminoethyl-5-naphthylamine-1-sulfonic acid sodium salt, 5-2-aminoethylamino-1-naphthalenesulphonic acid sodium salt, 1,5-edans, sodium salt, sodium 5-2-aminoethylamino-1-naphthalenesulfonate, sodium n-2-aminoethyl-5-naphthylamine-1-sulfonate, sodium5-2-aminoethylamino-1-naphthalenesulfonatehydrate
PubChem CID 4116252
IUPAC Name sodium;5-(2-aminoethylamino)naphthalene-1-sulfonate
SMILES C1=CC2=C(C=CC=C2S(=O)(=O)[O-])C(=C1)NCCN.[Na+]

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 4283850
Formula Weight 288.3
Percent Purity 97%
Chemical Name or Material 5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only