accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OBiONOOONOOONOO

CAS RN 10035-06-0

IUPAC Name: bismuth(3+) pentahydrate trinitrate
Synonyms: bismuth(III) nitrate
Molecular Weight (g/mol): 485.07
Molecular Formula: BiH10N3O14
InChi Key: FBXVOTBTGXARNA-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
  • (2)
  • (2)
  • (8)
  • (1)
  • (6)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (10)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (8)
  • (6)
  • (10)
  • (2)
  • (2)
  • (2)
  • (8)
  • (1)
  • (6)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (10)
  • (3)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)