missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth Nitrate, Pentahydrate, Crystal, ACS, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 50 kg |
| Percent Purity | 98% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Bismuth Nitrate, Pentahydrate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.