CAS RN 10034-96-5
Manganese Sulfate Monohydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese(II) sulfate monohydrate, 98+%, ACS reagent
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+);sulfate;hydrate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese(II) sulfate monohydrate, 99%
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+);sulfate;hydrate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese(II) sulfate monohydrate, 97%
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese(II) sulfate monohydrate, 99+%, extra pure
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+);sulfate;hydrate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganous Sulfate Solution, 364 g/L, Ricca Chemical
Molecular Formula: H2MnO5S MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganous Sulfate Monohydrate, ACS Reagent Grade, Ricca Chemical
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganous Sulfate, Monohydrate, ACS Grade, 98.0 to 101.0%, LabChem™
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganous Sulfate, MnSO4-H2O, MP Biomedicals
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate PubChem CID: 177577 ChEBI: CHEBI:86364 IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese Sulfate Monohydrate, Purified, Reagents
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L Synonym: Manganous Sulfate Monohydrate IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Manganese Sulfate, Monohydrate, Powder, USP, 98-102%, Spectrum™ Chemical
CAS: 10034-96-5 Molecular Formula: H2MnO5S Molecular Weight (g/mol): 169.01 MDL Number: MFCD00149159 InChI Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L IUPAC Name: manganese(2+) hydrate sulfate SMILES: O.[Mn++].[O-]S([O-])(=O)=O
- 1
- 2