missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals

Catalog No. ICN19470290
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19470290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19470290 Supplier MP Biomedicals Inc Supplier No. 0219470290

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crystalline grade of manganese sulfate monohydrate suitable for general laboratory research.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 169
Grade Cell Culture
Chemical Name or Material Manganese Sulfate Monohydrate
Recommended Storage Room Temperature