accessibility menu, dialog, popup

UserName

CAS RN 10030-80-5

IUPAC Name: (2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
Synonyms: L-mannose aldehydo-L-manno-hexose aldehydo-L-mannose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: GZCGUPFRVQAUEE-BXKVDMCESA-N
SMILES: OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)