missing translation for 'onlineSavingsMsg'
Learn More

Barium ICP Standard, 10,000 ppm Ba in 5% HNO3, Ricca Chemical

Catalog No. PBA10KN500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PBA10KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PBA10KN500 Supplier Ricca Chemical Company Supplier No. PBA10KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10022-31-8 , 7697-37-2 , 7732-18-5
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
PubChem CID 24798
IUPAC Name barium(2+) dinitrate
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Ba
CAS Min % 90.0, 6.73, 1.71
CAS Max % 93.0, 7.0, 1.78
Equivalence 1 mL = 10 mg Ba
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 500 mL
Chemical Name or Material Barium ICP Standard
Show More Show Less