missing translation for 'onlineSavingsMsg'
Learn More

Barium ICP Standard, 10,000 ppm Ba in 5% HNO3, Ricca Chemical

Catalog No. PBA10KN50
Change view
Click to view available options
Package: Natural Poly; Size: 50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PBA10KN50 Package: Natural Poly; Size: 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PBA10KN50 Supplier Ricca Chemical Company Supplier No. PBA10KN50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 10022-31-8, 7697-37-2, 7732-18-5
Concentration or Composition Notes 10,000ppm Ba
CAS Min % 90.0, 6.73, 1.71
CAS Max % 93.0, 7.0, 1.78
Equivalence 1 mL = 10 mg Ba
Molecular Formula BaN2O6
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name barium(2+) dinitrate
Molecular Weight (g/mol) 261.34
PubChem CID 24798
Chemical Name or Material Barium ICP Standard
Show More Show Less