missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium ICP Standard, 10,000 ppm Ba in 5% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS | 10022-31-8, 7697-37-2, 7732-18-5 |
| Concentration or Composition Notes | 10,000ppm Ba |
| CAS Min % | 90.0, 6.73, 1.71 |
| CAS Max % | 93.0, 7.0, 1.78 |
| Equivalence | 1 mL = 10 mg Ba |
| Molecular Formula | BaN2O6 |
| MDL Number | MFCD00003442 |
| InChI Key | IWOUKMZUPDVPGQ-UHFFFAOYSA-N |
| SMILES | [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Show More |