missing translation for 'onlineSavingsMsg'
Learn More

Barium ICP Standard, 1000 ppm Ba in 3% HNO3, Ricca Chemical

Catalog No. PBA1KN500
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PBA1KN500 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PBA1KN500 Supplier Ricca Chemical Company Supplier No. PBA1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 10022-31-8, 7697-37-2, 7732-18-5
Concentration or Composition Notes 1,000ppm Ba
CAS Min % 94.0, 4.22, 0.18
CAS Max % 97.0, 4.39, 0.19
Equivalence 1 mL = 1 mg Ba
Molecular Formula BaN2O6
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name barium(2+) dinitrate
Molecular Weight (g/mol) 261.34
PubChem CID 24798
Chemical Name or Material Barium ICP Standard
Show More Show Less