accessibility menu, dialog, popup

UserName

CAS RN 100-63-0

IUPAC Name: phenylhydrazine
Synonyms: phenylhydrazine|fenylhydrazine|fenilidrazina|ccris 511|phenylhydrazine (substance)
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: HKOOXMFOFWEVGF-UHFFFAOYSA-N
SMILES: NNC1=CC=CC=C1
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)