missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine, 97%

Catalog No. AAA1124630
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1124630 250 g
AAA1124618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1124630 Supplier Thermo Scientific Chemicals Supplier No. A1124630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-63-0 | C6H8N2 | 108.144 g/mol

Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

Solubility
Soluble in dilute acids.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 100-63-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007573
InChI Key HKOOXMFOFWEVGF-UHFFFAOYSA-N
Synonym hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
PubChem CID 7516
ChEBI CHEBI:27924
IUPAC Name phenylhydrazine
SMILES NNC1=CC=CC=C1

Specifications

Melting Point 18°C to 20°C
Density 1.099
Boiling Point 238°C to 241°C
Flash Point 88°C (190°F)
Assay Percent Range 97%
Refractive Index 1.6065
Quantity 250 g
UN Number UN2572
Beilstein 606080
Sensitivity Air and light sensitive
Merck Index 14,7293
Solubility Information Soluble in dilute acids.
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material Phenylhydrazine
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H317-H319-H335-H341-H350-H372
Hazard Statement GHS H Statement
H301-H311-H331-H350-H372-H341-H227-H315-H319-H317 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENYLHYDRAZINE
EINECSNumber 202-873-5
RTECSNumber MV8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only