missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ ZWITTERGENT™ 3-14 Detergent, Calbiochem™,
$242.00 - $4,300.00
Chemical Identifiers
| CAS | 14933-09-6 |
|---|---|
| Molecular Formula | C19H41NO3S |
| Molecular Weight (g/mol) | 363.601 |
| InChI Key | BHATUINFZWUDIX-UHFFFAOYSA-N |
| Synonym | n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
| PubChem CID | 84705 |
| ChEBI | CHEBI:10129 |
| IUPAC Name | 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate |
| SMILES | CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
6930175GM
|
MilliporeSigma™
6930175GM |
5 g | Glass Bottle |
Each for $242.00
|
|
||||
|
69301725GM
|
MilliporeSigma™
69301725GM |
25 g | Glass Bottle |
Each for $756.00
|
|
||||
|
693017100GM
|
MilliporeSigma™
693017100GM |
100 g | Glass Bottle |
Each for $2,650.00
|
|
||||
|
693017500GM
|
MilliporeSigma™
693017500GM |
500 g | Glass Bottle |
Each for $4,300.00
|
|
||||
Chemical Identifiers
| 14933-09-6 | |
| 363.601 | |
| n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate | |
| CHEBI:10129 | |
| CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| C19H41NO3S | |
| BHATUINFZWUDIX-UHFFFAOYSA-N | |
| 84705 | |
| 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate |
Specifications
| White | |
| C19H41NO3S | |
| BHATUINFZWUDIX-UHFFFAOYSA-N | |
| 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate | |
| 84705 | |
| ≥99% (TLC) |
| 14933-09-6 | |
| n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate | |
| CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
| 363.601 | |
| CHEBI:10129 |