missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZWITTERGENT™ 3-14 Detergent, Calbiochem™,

Catalog No. 6930175GM
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6930175GM 5 g Glass Bottle
69301725GM 25 g Glass Bottle
693017100GM 100 g Glass Bottle
693017500GM 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6930175GM Supplier MilliporeSigma™ Supplier No. 6930175GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14933-09-6
Molecular Formula C19H41NO3S
Molecular Weight (g/mol) 363.601
InChI Key BHATUINFZWUDIX-UHFFFAOYSA-N
Synonym n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 84705
ChEBI CHEBI:10129
IUPAC Name 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥99% (TLC)