missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZWITTERGENT™ 3-14 Detergent, Calbiochem™,

Supplier:  MilliporeSigma™ 69301725GM

Quantity 25 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. 69301725GM


Only null left

Description

Description

Specifications

Chemical Identifiers

14933-09-6
363.601
n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
CHEBI:10129
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
C19H41NO3S
BHATUINFZWUDIX-UHFFFAOYSA-N
84705
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate

Specifications

White
25 g
C19H41NO3S
BHATUINFZWUDIX-UHFFFAOYSA-N
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
84705
≥99% (TLC)
14933-09-6
Glass Bottle
n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
363.601
CHEBI:10129
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions