missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ ZWITTERGENT™ 3-14 Detergent, Calbiochem™,
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Packaging | Glass Bottle |
| Percent Purity | ≥99% (TLC) |
| Color | White |
| Quantity | 500 g |
| Packaging | Glass Bottle |
| Percent Purity | ≥99% (TLC) |